Learn More
Fisherbrand™ Borosilicate Glass Narrow Neck Erlenmeyer Flasks
Constructed of borosilicate glass. Fisherbrand™ Borosilicate Glass Narrow Neck Erlenmeyer Flasks are conical and graduated.
Brand:  Fisherbrand™ SPFS66500-5000
53.24 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 15479103
  • 112.00€ / Each
In Stock
to see stock.
Edge
Description
  • Graduated
  • ISO 1773
  • Borosilicate glass
  • Conical
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 1773
5 L
365 mm
Yes
220 mm
50 mm
Borosilicate Glass
Conical
Narrow
SDS
Documents Certificates
Special Offers
Special Offers
  • Fisherbrand
    Get up to 60% off select Fisherbrand products
    Promo Code: 24162
    Expires2025-03-31
    Days
    0
    Hours
    10
    Minutes
    33
    Seconds
    45
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions Save Now
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    29
    Hours
    13
    Minutes
    26
    Seconds
    14
  • Multiple Manufacturers
    Explore exclusive monthly deals on essential consumables for unbeatable savings.
    Promo Code: 24222
    Expires2025-03-31
    Days
    0
    Hours
    10
    Minutes
    33
    Seconds
    45
  • Multiple Manufacturers
    Browse exclusive prices for our academic customers
    Promo Code: 25081
    Expires2025-12-31
Product Content Correction
Product Title
Fisherbrand™ Borosilicate Glass Narrow Neck Erlenmeyer Flasks > Capacity: 5000mL