Learn More
Witeg™ Brass Cork Borer Sets
Use the Witeg™ Brass Cork Borer Set for your tooling needs.
306.00€ - 1035.00€
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | ||||||
11807780
|
Witeg™
7 414 303 |
5 to 18.75 mm |
626.00€
Each |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
11887770
|
Witeg™
7 414 301 |
5 to 11 mm |
306.00€
Each |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
11897770
|
Witeg™
7 414 305 |
5 to 26.25 mm |
1035.00€
Each |
Estimated Shipment: 07-07-2025
Log in to see stock. |
||||||
Description
- Cork
- Brass
Specifications
Bork Borer Set | |
Brass |
Product Title
Witeg™ Brass Cork Borer Sets