Learn More
Witeg™ Brass Cork Borer Sets
Use the Witeg™ Brass Cork Borer Set for your tooling needs.
Brand:  Witeg™ 7 414 303
 View more versions of this product
Product Code. 11807780
  • 626.00€ / Each
Estimated Shipment: 16-04-2025
Description
  • Cork
  • Brass
Specifications
Specifications
Bork Borer Set
5 to 18.75 mm
Brass
12Pack
Product Title
Witeg™ Brass Cork Borer Sets > Pusher Diameter: 5 to 18.75mm