Learn More
WTW™ Turb™ Calibration Kits
Get optimum calibration results for your turbidimeters with the Turb™ calibration kits.
Brand: WTW™ 600547
331.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Includes Cleaning tissue and indexing rings
- For use with Turb® 355 T, 335 IR Portable turbidimeter
- Turbidity of 0.02 - 10 - 1000NTU
- Calibration set
Specifications
Turb 355 T, 335 IR Portable Turbidimeter | |
Calibration set | |
Cleaning tissue and indexing rings | |
0.02, 10, 1000 NTU |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
WTW™ Turb™ Calibration Kits > For use with Turb 355 Series Portable turbidimeter