missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ BTG1 (Human) Recombinant Protein
Human BTG1 full-length ORF ( AAH16759, 1 a.a. - 171 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000694-P01.10ug
This item is not returnable.
View return policy
Description
- Sequence: MHPFYTRAATMIGEIAAAVSFISKFLRTKGLTSERQLQTFSQSLQELLAEHYKHHWFPEKPCKGSGYRCIRINHKMDPLIGQAAQRIGLSSQELFRLLPSELTLWVDPYEVSYRIGEDGSICVLYEASPAGGSTQNSTNVQMVDSRISCKEELLLGRTSPSKNYNMMTVSG
Specifications
AAH16759 | |
B-cell translocation gene 1, anti-proliferative | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
694 | |
Wheat germ expression system | |
10 μg | |
BTG1 | |
GST |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction