Learn More
Uvex Busingess Blue PU 84262 Boot - Width 11
Uvex 1 shoes designed for classic business look while providing protection against workplace risks; extra-wide steel toecap, penetration-resistant midsole and SRC polyurethane outsole protects toes and keeps from slipping; ideal for active men in office and factory environments
139.00€
Specifications
Color Blue
Material Microvelour
Sole Material Polyurethane (PU)
Toe Material Steel
Disposable Reusable
View More Specs
Products 14
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17284451
View Documents Promotion Details
Uvex
8426239
39
139.00€
1 pair
Estimated Shipment: 22-11-2024
to see stock.
Add to basket
17294451
View Documents Promotion Details
Uvex
8426240
40
139.00€
1 pair
Estimated Shipment: 22-11-2024
to see stock.
Add to basket
17204461
View Documents Promotion Details
Uvex
8426241
41
139.00€
1 pair
Estimated Shipment: 22-11-2024
to see stock.
Add to basket
17214461
View Documents Promotion Details
Uvex
8426242
42
139.00€
1 pair
Estimated Shipment: 22-11-2024
to see stock.
Add to basket
17224461
View Documents Promotion Details
Uvex
8426243
43
139.00€
1 pair
Estimated Shipment: 22-11-2024
to see stock.
Add to basket
17234461
View Documents Promotion Details
Uvex
8426244
44
139.00€
1 pair
Estimated Shipment: 22-11-2024
to see stock.
Add to basket
17244461
View Documents Promotion Details
Uvex
8426245
45
139.00€
1 pair
Estimated Shipment: 22-11-2024
to see stock.
Add to basket
17254461
View Documents Promotion Details
Uvex
8426246
46
139.00€
1 pair
Estimated Shipment: 22-11-2024
to see stock.
Add to basket
17264461
View Documents Promotion Details
Uvex
8426247
47
139.00€
1 pair
Estimated Shipment: 22-11-2024
to see stock.
Add to basket
17274461
View Documents Promotion Details
Uvex
8426248
48
139.00€
1 pair
Estimated Shipment: 22-11-2024
to see stock.
Add to basket
17284461
View Documents Promotion Details
Uvex
8426249
49
139.00€
1 pair
Estimated Shipment: 22-11-2024
to see stock.
Add to basket
17294461
View Documents Promotion Details
Uvex
8426250
50
139.00€
1 pair
Estimated Shipment: 22-11-2024
to see stock.
Add to basket
17204471
View Documents Promotion Details
Uvex
8426251
51
139.00€
1 pair
Estimated Shipment: 22-11-2024
to see stock.
Add to basket
17214471
View Documents Promotion Details
Uvex
8426252
52
139.00€
1 pair
Estimated Shipment: 22-11-2024
to see stock.
Add to basket
Description
Description
Uvex 1 business Chelsea Boot 8426.2 S3 SRC
  • Safe, smart, stylish
  • Fashionably business look with trendy design
  • Generous fit with wide steel toe cap
  • Penetration-resistant, non-metallic midsole in accordance with latest safety standards, does not restrict flexibility
  • Uvex Climazone technology ensures optimum wearer comfort
  • Breathable, water-resistant high-tech microvelour upper material
  • Textile lining provides optimum breathability
  • Made from synthetic materials, suitable for people allergic to chrome
  • Shock absorbing, durable mono density polyurethane sole unit
  • Removable comfort insole, antistatic with moisture control system and shock absorption in heel and at front of foot
  • Sole unit of free of lacquer wetting inhibitors such as silicones, phthalates, etc.
  • Fulfills ESD requirements, resistance to ground Kleiner 35 megohms
  • Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
  • Article No. 8426.2
  • Standard EN ISO 20345:2011 S3 SRC
  • Sizes: 39 to 52
Specifications
Specifications
Blue
Polyurethane (PU)
Reusable
Men's
Uvex 1
No
SRC
Microvelour
Steel
EN ISO 20345:2011
Standard Width
Yes
S3