Learn More
DiscoverX cAMP Hunter™ CHO-K1 AGTRL1 Gi Cell Line
The cAMP Hunter™ CHO-K1 AGTRL1 Gi Cell Line measures AGTRL1 (GPCR) activity via decreased levels of intracellular cAMP. This cell line ships as two vials of cryopreserved cells.
Brand: DiscoverX 95-0147C2
This item is not returnable.
View return policy
Description
The cAMP Hunter AGTRL1 Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. apelin-13) based activation of AGTRL1 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype AGTRL1, a naturally Gi/Go coupled GPCR. Upon activation of AGTRL1, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells.Specifications
Intracellular cAMP level | |
Cell Lines | |
2 Vials | |
NM_005161 |
CHO-K1 | |
Apelin-13 trifluoroacetate salt | |
AGTRL1 |