Learn More
DiscoverX cAMP Hunter™ CHO-K1 GALR1 Gi Cell Line
The cAMP Hunter™ CHO-K1 GALR1 Gi Cell Line measures GALR1 (GPCR) activity via decreased levels of intracellular cAMP. This cell line ships as two vials of cryopreserved cells.
Brand:  DiscoverX 95-0058C2
Encompass
Product Code. 17304489
  • 19183.50€ / 2 ampoules
Estimated Shipment: 18-02-2025
Description
The cAMP Hunter GALR1 Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. galanin) based activation of GALR1 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype GALR1 , a naturally Gi/Go coupled GPCR. Upon activation of GALR1 , the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
Intracellular cAMP level
Cell Lines
2 Vials
AY541036
CHO-K1
Galanin
GALR1
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.