Learn More
DiscoverX cAMP Hunter™ CHO-K1 GALR1 Gi Cell Line
The cAMP Hunter™ CHO-K1 GALR1 Gi Cell Line measures GALR1 (GPCR) activity via decreased levels of intracellular cAMP. This cell line ships as two vials of cryopreserved cells.
Brand: DiscoverX 95-0058C2
This item is not returnable.
View return policy
Description
The cAMP Hunter GALR1 Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. galanin) based activation of GALR1 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype GALR1 , a naturally Gi/Go coupled GPCR. Upon activation of GALR1 , the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells.Specifications
Intracellular cAMP level | |
Cell Lines | |
2 Vials | |
AY541036 |
CHO-K1 | |
Galanin | |
GALR1 |