Learn More
DiscoverX cAMP Hunter™ CHO-K1 HTR1A Gi Cell Line
The cAMP Hunter™ CHO-K1 HTR1A Gi Cell Line measures HTR1A (GPCR) activity via decreased levels of intracellular cAMP. This cell line ships as two vials of cryopreserved cells.
Brand: DiscoverX 95-0132C2
Description
The cAMP Hunter HTR1A Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin/5-HT) based activation of HTR1A GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype HTR1A , a naturally Gi/Go coupled GPCR. Upon activation of HTR1A , the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells.Specifications
Intracellular cAMP level | |
Cell Lines | |
2 Vials | |
NM_000524.2 |
CHO-K1 | |
Serotonin/5-HT | |
HTR1A |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.