Learn More
DiscoverX cAMP Hunter™ CHO-K1 NPBWR1 Gi Cell Line
The cAMP Hunter™ CHO-K1 NPBWR1 Gi Cell Line measures NPBWR1 (GPCR) activity via decreased levels of intracellular cAMP. This cell line ships as two vials of cryopreserved cells.
Brand: DiscoverX 95-0099C2
This item is not returnable.
View return policy
Description
The cAMP Hunter NPBWR1 Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. neuropeptides B and W) based activation of NPBWR1 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype NPBWR1, a naturally Gi/Go coupled GPCR. Upon activation of NPBWR1, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells.Specifications
Intracellular cAMP level | |
2 vials | |
Neuropeptide W-23 | |
NPBWR1 |
CHO-K1 | |
Cell Lines | |
2 Vials | |
NM_005285 |