Learn More
DiscoverX cAMP Hunter™ CHO-K1 OPRD1 Gi Cell Line
The cAMP Hunter™ CHO-K1 OPRD1 Gi Cell Line measures OPRD1 (GPCR) activity via decreased levels of intracellular cAMP. This cell line ships as two vials of cryopreserved cells.
Brand:  DiscoverX 95-0108C2
Encompass
Product Code. 17304609
  • 19183.50€ / 2 ampoules
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The cAMP Hunter OPRD1 Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. DADLE, DPDPE) based activation of OPRD1 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype OPRD1, a naturally Gi/Go coupled GPCR. Upon activation of OPRD1, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
Intracellular cAMP level
Cell Lines
2 Vials
NM_000911
CHO-K1
[D-Ala2, D-Leu5]-Enkephalin (DADLE)
OPRD1