Learn More
DiscoverX cAMP Hunter™ CHO-K1 PROKR1 Gs Cell Line
The cAMP Hunter™ CHO-K1 PROKR1 Gs Cell Line measures PROKR1 (GPCR) activity via increased levels of intracellular cAMP. This cell line ships as two vials of cryopreserved cells.
Brand:  DiscoverX 95-0090C2
Encompass
Product Code. 17304569
  • 19183.50€ / 2 ampoules
Estimated Shipment: 05-02-2025
Description
The cAMP Hunter PROKR1 Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prokinectin, EG-VEGF) based activation of PROKR1 GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype PROKR1, a naturally Gs coupled GPCR. Upon activation of PROKR1, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
Intracellular cAMP level
Cell Lines
2 Vials
NM_138964
CHO-K1
Recombinant Human EG-VEGF
PROKR1
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.