Learn More
DiscoverX cAMP Hunter™ CHO-K1 TSHR (Long Isoform) Gs Cell Line
The cAMP Hunter™ CHO-K1 TSHR (Long Isoform) Gs Cell Line measures TSHRL (GPCR) activity via increased levels of intracellular cAMP. This cell line ships as two vials of cryopreserved cells.
Brand:  DiscoverX 95-0153C2
Encompass
Product Code. 17344689
  • 14968.50€ / 2 ampoules
Estimated Shipment: 17-01-2025
Description
The cAMP Hunter TSHRL Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. TSH) based activation of TSHRL GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype TSHRL , a naturally Gs coupled GPCR. Upon activation of TSHRL , the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
Intracellular cAMP level
Cell Lines
2 Vials
NM_000369.2
CHO-K1
TSH, Thyroid stimulating hormone, Human
TSHRL
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.