Learn More
Thermo Scientific™ CaptureSelect™ GCSFSelect Leakage ELISA
Thermo Scientific™ CaptureSelect™ GCSFSelect Leakage ELISA is designed to detect possible leached ligand of human GCSF purified using GCSFSelect affinity resin (Cytiva).
1065.00€ - 9240.00€
Specifications
Content And Storage | Store at -5°C to -30°C. |
---|---|
Assay | ELISA |
For Use With (Equipment) | Microplate Reader |
Product Line | CaptureSelect™ |
Target | GCSFSelect Affinity Ligand |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18486638
|
Thermo Scientific™
810313001 |
1 Assay |
1065.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18406608
|
Thermo Scientific™
810313010 |
10 Assays |
9240.00€
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CaptureSelect GCSFSelect Leakage ELISA is designed to detect possible leached ligand of human GCSF purified using GCSFSelect affinity resin (Cytiva). Even when covalently attached, small amounts of the IgSelect affinity ligand can leach off of the chromatography support and co-elute with the antibody.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect GCSFSelect Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
Specifications
Store at -5°C to -30°C. | |
Microplate Reader | |
GCSFSelect Affinity Ligand | |
Kit |
ELISA | |
CaptureSelect™ | |
ELISA Kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.