Learn More
Thermo Scientific™ CaptureSelect™ HSA Ligand Leakage ELISA
Thermo Scientific™ CaptureSelect™ HSA Ligand Leakage ELISA is designed to detect possible leached ligand of human serum albumin (HSA) or HSA fusion products purified using CaptureSelect Human Albumin affinity matrix.
1060.00€ - 9240.00€
Specifications
Content And Storage | Store in freezer (-5°C to -30°C). |
---|---|
Product Line | CaptureSelect™ |
Type | ELISA Kit |
Format | Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18496498
|
Thermo Scientific™
810297001 |
1 Assay |
1060.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18456578
|
Thermo Scientific™
810297010 |
10 Assays |
9240.00€
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CaptureSelect HSA Ligand Leakage ELISA is designed to detect possible leached ligand of human serum albumin (HSA) or HSA fusion products purified using the CaptureSelect Human Albumin affinity matrix. The HSA affinity ligand, a 13 kD recombinant protein, is coupled by multipoint attachment through stable amide bonds to give high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The HSA Ligand Leakage ELISA kit can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
Specifications
Store in freezer (-5°C to -30°C). | |
CaptureSelect™ | |
ELISA Kit | |
Kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.