Learn More
Thermo Scientific™ CaptureSelect™ VIII Select Leakage ELISA
Thermo Scientific™ CaptureSelect™ VIIISelect Leakage ELISA is designed to detect possible leached ligand of B domain-depleted recombinant Factor VIII purified using VIIISelect affinity resin (Cytiva).
1065.00€ - 9240.00€
Specifications
Content And Storage | Store at -5°C to -30°C. |
---|---|
Assay | ELISA |
For Use With (Equipment) | Microplate Reader |
Product Line | CaptureSelect™ |
Target | VIIISelect Affinity Ligand |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18476738
|
Thermo Scientific™
810286001 |
1 Assay |
1065.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18466478
|
Thermo Scientific™
810286010 |
10 Assays |
9240.00€
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CaptureSelect VIIISelect Leakage ELISA is designed to detect possible leached ligand of B domain-depleted recombinant Factor VIII purified using VIIISelect affinity resin (Cytiva). Even when covalently attached, small amounts of the VIIISelect affinity ligand can leach off of the chromatography support and co-elute with human Factor VIII.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect VIIISelect Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
Specifications
Store at -5°C to -30°C. | |
Microplate Reader | |
VIIISelect Affinity Ligand | |
Kit |
ELISA | |
CaptureSelect™ | |
ELISA Kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.