missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Mouse anti-Human , BUV615, Clone: SJ25C1 (also known as SJ25-C1), BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 612990
This item is not returnable.
View return policy
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow CytometrySpecifications
CD19 | |
Monoclonal | |
Brilliant Ultraviolet 615 | |
B4; B-lymphocyte antigen CD19; Leu-12 | |
Human NALM1 + NALM16 cells | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
SJ25C1 (also known as SJ25-C1) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |