missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Mouse anti-Human , BUV615, Clone: SJ25C1 (also known as SJ25-C1), BD Biosciences™
Mouse Monoclonal Antibody
378.00€ - 750.00€
Specifications
Antigen | CD19 |
---|---|
Clone | SJ25C1 (also known as SJ25-C1) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Ultraviolet 615 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17218034
|
BD Biosciences
612990 |
25 Tests |
378.00€
25 tests |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
17208034
|
BD Biosciences
612989 |
100 Tests |
750.00€
100 tests |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow CytometrySpecifications
CD19 | |
Flow Cytometry | |
Brilliant Ultraviolet 615 | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human NALM1 + NALM16 cells | |
Primary |
SJ25C1 (also known as SJ25-C1) | |
Monoclonal | |
Mouse | |
Human | |
B4; B-lymphocyte antigen CD19; Leu-12 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |