missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse anti-Human, BUV805, Clone: UCHT1, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 612896
205.16 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow Cytometry.
Specifications
CD3 | |
Monoclonal | |
Brilliant Ultraviolet 805 | |
25 Tests | |
Primary | |
Human | |
IgG1 κ |
Flow Cytometry | |
UCHT1 | |
Mouse | |
RUO | |
CD3 | |
Refrigerate (2 to 8°C) |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD3 Mouse anti-Human, BUV805, Clone: UCHT1, BD Horizon™ > 25 Tests; BUV805
Spot an opportunity for improvement?Share a Content Correction