missing translation for 'onlineSavingsMsg'
Learn More

CD3 Mouse anti-Human, BUV805, Clone: UCHT1, BD Horizon™

Mouse Monoclonal Antibody

206.00€ - 510.00€

Specifications

Antigen CD3
Clone UCHT1
Applications Flow Cytometry
Classification Monoclonal
Conjugate Brilliant Ultraviolet 805
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17207954
View Documents
BD Biosciences
612896
25 Tests
206.00€
25 tests
Please to purchase this item. Need a web account? Register with us today!
17297944
View Documents
BD Biosciences
612895
100 Tests
510.00€
100 tests
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD3
Flow Cytometry
Brilliant Ultraviolet 805
RUO
IgG1 κ
Refrigerate (2 to 8°C)
UCHT1
Monoclonal
Mouse
Human
Primary
CD3
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CD3 Mouse anti-Human, BUV805, Clone: UCHT1, BD Horizon™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.