Learn More
CD4 Mouse, anti-Human, R718, Clone: SK3, BD Horizon™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 566930
 View more versions of this product
Product Code. 17297714
  • 422.00€ / 100 tests
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The SK3 monoclonal antibody specifically binds to CD4. The CD4 antigen is a 55 kDa type 1 transmembrane glycoprotein that belongs to the immunoglobulin superfamily. It is expressed on T-helper/inducer lymphocytes and monocytes. CD3+CD4+ T cells comprise approximately 28% to 58% of normal peripheral blood lymphocytes. CD4 is also expressed on 80% to 95% of normal thymocytes. The CD4 antigen is present in low density on the cell surface of monocytes and in the cytoplasm of monocytes and macrophages (CD3-CD4+). CD4 serves as a T cell coreceptor for MHC class II antigen recognition and as a receptor for the human immunodeficiency virus (HIV). The SK3 antibody inhibits HIV binding to CD4+ cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD4
Monoclonal
R718
100 Tests
Primary
Refrigerate Upon Arrival
Flow Cytometry
SK3