Learn More
CD4 Mouse, anti-Human, R718, Clone: SK3, BD Horizon™
Mouse Monoclonal Antibody
183.00€ - 422.00€
Specifications
Antigen | CD4 |
---|---|
Clone | SK3 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | R718 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17207724
|
BD Biosciences
566931 |
25 Tests |
183.00€
25 tests |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
17297714
|
BD Biosciences
566930 |
100 Tests |
422.00€
100 tests |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
Description
The SK3 monoclonal antibody specifically binds to CD4. The CD4 antigen is a 55 kDa type 1 transmembrane glycoprotein that belongs to the immunoglobulin superfamily. It is expressed on T-helper/inducer lymphocytes and monocytes. CD3+CD4+ T cells comprise approximately 28% to 58% of normal peripheral blood lymphocytes. CD4 is also expressed on 80% to 95% of normal thymocytes. The CD4 antigen is present in low density on the cell surface of monocytes and in the cytoplasm of monocytes and macrophages (CD3-CD4+). CD4 serves as a T cell coreceptor for MHC class II antigen recognition and as a receptor for the human immunodeficiency virus (HIV). The SK3 antibody inhibits HIV binding to CD4+ cells.
Specifications
CD4 | |
Flow Cytometry | |
R718 | |
Mouse | |
Human | |
Refrigerate Upon Arrival |
SK3 | |
Monoclonal | |
Liquid | |
RUO | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.