Learn More
CD4 Mouse, anti-Human, R718, Clone: SK3, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566931
Description
The SK3 monoclonal antibody specifically binds to CD4. The CD4 antigen is a 55 kDa type 1 transmembrane glycoprotein that belongs to the immunoglobulin superfamily. It is expressed on T-helper/inducer lymphocytes and monocytes. CD3+CD4+ T cells comprise approximately 28% to 58% of normal peripheral blood lymphocytes. CD4 is also expressed on 80% to 95% of normal thymocytes. The CD4 antigen is present in low density on the cell surface of monocytes and in the cytoplasm of monocytes and macrophages (CD3-CD4+). CD4 serves as a T cell coreceptor for MHC class II antigen recognition and as a receptor for the human immunodeficiency virus (HIV). The SK3 antibody inhibits HIV binding to CD4+ cells.
Specifications
CD4 | |
Monoclonal | |
R718 | |
25 Tests | |
Primary | |
Refrigerate Upon Arrival |
Flow Cytometry | |
SK3 | |
Mouse | |
RUO | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.