missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Mouse anti-Human , PE-Cyanine7, Clone: HIT8a, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566859
This item is not returnable.
View return policy
Description
CD8 Monoclonal antibody specifically detects CD8 in Human samples. It is validated for Flow CytometrySpecifications
CD8 | |
Monoclonal | |
PE-Cyanine7 | |
CD8α; CD8A; CD8 alpha; Leu2a; MAL; T8; p32 | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with PE-Cyanine7 under optimum conditions, and unconjugated antibody and free PE-Cyanine7 were removed. |
Flow Cytometry | |
HIT8a | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
RUO | |
Human | |
IgG1 κ |