missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Mouse anti-Human , PE-Cyanine7, Clone: HIT8a, BD Biosciences™
Mouse Monoclonal Antibody
139.00€ - 452.00€
Specifications
Antigen | CD8 |
---|---|
Clone | HIT8a |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE-Cyanine7 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17217664
|
BD Biosciences
566859 |
25 Tests |
139.00€
25 tests |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
17207664
|
BD Biosciences
566858 |
100 Tests |
452.00€
100 tests |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
Description
CD8 Monoclonal antibody specifically detects CD8 in Human samples. It is validated for Flow CytometrySpecifications
CD8 | |
Flow Cytometry | |
PE-Cyanine7 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with PE-Cyanine7 under optimum conditions, and unconjugated antibody and free PE-Cyanine7 were removed. |
HIT8a | |
Monoclonal | |
Mouse | |
Human | |
CD8α; CD8A; CD8 alpha; Leu2a; MAL; T8; p32 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title