Learn More
CDCP1 Antibody (309121), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB26662J
Description
CDCP1 Monoclonal antibody specifically detects CDCP1 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, CyTOF
Specifications
CDCP1 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
64866 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, Immunoprecipitation, CyTOF | |
309121 | |
Flow Cytometry, Immunoprecipitation, CyTOF-ready | |
CD318, CD318 antigen, CUB domain containing protein 1, CUB domain-containing protein 1, Membrane glycoprotein gp140, SIMA135TRASK, Subtractive immunization M plus HEp3-associated 135 kDa protein, Transmembrane and associated with src kinases | |
Mouse myeloma cell line NS0-derived recombinant human CDCP1 isoform 1, Ala33-Leu666, Accession # NP_073753 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.