Learn More
Greiner Bio-One CELLSTAR ÎĽClear™ 96-well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate
GREENER_CHOICE
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
Brand:  Greiner Bio-One 675090
Product Code. 10762394
  • 281.20€ / Pack of 32
Only null left
Estimated Shipment: 06-08-2025
Add to basket
Description
Description
Alphanumeric coding
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Half-Area
8 x 12
Black, Clear Bottom
96
Sterile
199 ÎĽL
Yes
32
Yes
127.76 x 85.48 x 14.40 mm
With Lid
Not Autoclavable
Polystyrene
15 to 175 ÎĽL
Cell Culture
Round, Flat Bottom (F-well)
4800 x g (swinging bucket rotor)
8
Black
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Greiner Bio-One CELLSTAR ÎĽClear™ 96-well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate > Black w/Clear Bottom