missing translation for 'onlineSavingsMsg'
Learn More
Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate Product Code.: 10501853
Add to basket

Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate
GREENER_CHOICE

Product Code. 10501853 Shop All Corning Products
20/Pk., 100/Cs.
Click to view available options
Packaging:
20/Pk., 100/Cs.
4/Pk
This item is not returnable. View return policy

Product Code. 10501853

Brand: Corning™ 3882

Add to basket
Add to basket
This item is not returnable. View return policy

Color: Black, Clear Bottom; Sterility: Sterile; Lid: With Lid

  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • Greener Choice Claims
    • Environmental benefits include:
      • Renewable Energy
      • Certifications and Seals of Approval
      • Energy Star Certification
    TRUSTED_SUSTAINABILITY

    Specifications

    Format Half-Area
    Lid With Lid
    Culture Area 0.16 cm2
    Array 8 x 12
    Packaging 20/Pk., 100/Cs.
    Color Black, Clear Bottom
    Product Type Half Area
    For Use With (Equipment) Standard automated handling and detection systems
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 25 to 125 μL
    Sterility Sterile
    Surface Treatment Cell Culture
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Homogenous and cell based assays
    No. per Case 100/Cs.
    No. per Pack 20/Pk.
    Quantity 100/Cs.
    Well Color Clear
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate >

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.