Learn More
DiscoverX ChemiScreen™ AT2 Angiotensin II Receptor Membrane Preparation
The ChemiScreen™ AT2 Angiotensin II Receptor Membrane Preparation allows measurement of ligand binding to AGTR2 GPCR via radioligand binding assays. Membrane preps contain 1mL of 1mg/mL frozen membranes.
Brand: DiscoverX HTS208M
This item is not returnable.
View return policy
Description
The ChemiScreen™ AT2 Angiotensin II Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of AGTR2 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1mL of 1mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression.Specifications
AT2 | |
Membrane Preps | |
1 Vial (200 units) | |
AGTR2 |