Learn More
DiscoverX ChemiScreen™ BB1 Bombesin/Neuromedin B Receptor Membrane Preparation
The ChemiScreen™ BB1 Bombesin/Neuromedin B Receptor Membrane Preparation allows measurement of ligand binding to NMBR GPCR via radioligand binding assays, or GPCR activity via GTPγS assay. Membrane preps contain 1mL of 1mg/mL frozen membranes.
Brand: DiscoverX HTS123M
This item is not returnable.
View return policy
Description
The ChemiScreen™ BB1 Bombesin/Neuromedin B Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of NMBR surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach or to screen for GPCR activity using GTPγS assays. Increased binding affinities or GPCR activation levels will lead to higher radioactive signal. Membrane preps contain 1mL of 1mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression.Specifications
BB1 | |
Membrane Preps | |
1 Vial (200 units) | |
NMBR |