Learn More
DiscoverX ChemiScreen™ CB1 Cannabinoid Receptor Membrane Preparation
The ChemiScreen™ CB1 Cannabinoid Receptor Membrane Preparation allows measurement of ligand binding to CNR1 GPCR via radioligand binding assays, or GPCR activity via GTPγS assay. Membrane preps contain 1mL of 1mg/mL frozen membranes.
Brand:  DiscoverX HTS019M
Encompass
Product Code. 17384989
  • 1048.50€ / 1 ampoule
Explore more special offers
Description
Description
The ChemiScreen™ CB1 Cannabinoid Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of CNR1 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach or to screen for GPCR activity using GTPγS assays. Increased binding affinities or GPCR activation levels will lead to higher radioactive signal. Membrane preps contain 1mL of 1mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression.
Specifications
Specifications
CB1
Membrane Preps
1 Vial
CNR1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
DiscoverX ChemiScreen™ CB1 Cannabinoid Receptor Membrane Preparation