Learn More
DiscoverX ChemiScreen™ CCR6 Receptor Chemokine Membrane Preparation
The ChemiScreen™ CCR6 Receptor Chemokine Membrane Preparation allows measurement of ligand binding to CCR6 GPCR via radioligand binding assays. Membrane preps contain 1mL of 1mg/mL frozen membranes.
Brand: DiscoverX HTS011M
This item is not returnable.
View return policy
Description
The ChemiScreen™ CCR6 Receptor Chemokine Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of CCR6 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1mL of 1mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression.Specifications
CCR6 | |
Membrane Preps | |
1 Vial | |
CCR6 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.