Learn More
DiscoverX ChemiScreen™ GPR68/OGR1 Proton-Sensing Receptor Stable Cell Line
The ChemiScreen™ GPR68/OGR1 Proton-Sensing Receptor Stable Cell Line measures GPR68 (GPCR) activation via an increase in intracellular calcium. This cell line ships as two vials of cryopreserved cells.
Brand: DiscoverX HTS153C
This item is not returnable.
View return policy
Description
The ChemiScreen™ GPR68/OGR1 Proton-Sensing Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of pH based activation of GPR68 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses GPR68 alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of GPR68 , the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells.Specifications
Calcium Flux | |
Cell Lines | |
2 Vials | |
NM_003485 |
Chem-1 | |
CHECK | |
GPR68 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
DiscoverX ChemiScreen™ GPR68/OGR1 Proton-Sensing Receptor Stable Cell Line