Learn More
DiscoverX ChemiScreen™ MC5 Melanocortin Receptor Membrane Preparation
The ChemiScreen™ MC5 Melanocortin Receptor Membrane Preparation allows measurement of ligand binding to MC5R GPCR via radioligand binding assays. Membrane preps contain 1mL of 1mg/mL frozen membranes.
Brand: DiscoverX HTS155M
This item is not returnable.
View return policy
Description
The ChemiScreen™ MC5 Melanocortin Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of MC5R surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1mL of 1mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression.Specifications
MC5 | |
Membrane Preps | |
1 Vial (200 units) | |
MC5R |