Learn More
DiscoverX ChemiScreen™ PAC1-long Receptor Stable Cell Line
The ChemiScreen™ PAC1-long Receptor Stable Cell Line measures ADCYAP1R1 (GPCR) activation via an increase in intracellular calcium. This cell line ships as two vials of cryopreserved cells.
Brand: DiscoverX HTS114C
This item is not returnable.
View return policy
Description
The ChemiScreen™ PAC1-long Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. PACAP) based activation of ADCYAP1R1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses ADCYAP1R1 alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of ADCYAP1R1 , the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells.Specifications
Calcium Flux | |
Cell Lines | |
2 Vials | |
AY366498 |
Chem-1 | |
PACAP (1-27) | |
ADCYAP1R1 |