Learn More
DiscoverX CHO-K1 PTAFR Gq Cell Line
The CHO-K1 PTAFR Gq Cell Line measures PTAFR (GPCR) activation via an increase in intracellular calcium. This cell line ships as two vials of cryopreserved cells.
Brand:  DiscoverX 95-0091C2
Encompass
Product Code. 17334569
  • 19183.50€ / 2 ampoules
Estimated Shipment: 28-01-2025
Description
The PTAFR Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. PAF) based activation of PTAFR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses PTAFR , a naturally Gq coupled GPCR. Upon activation of PTAFR , the resulting calcium activity can be fluorescently assayed using the Calcium NoWash PLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
Intracellular cAMP level
Cell Lines
2 Vials
NM_000952.4
CHO-K1
PAF
PTAFR
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.