Learn More
Witeg™ Class AS Special Sizes Graduated Pipettes
With tolerances according to DIN EN ISO 648.
Brand: Witeg™ 3 535 025
This item is not returnable.
View return policy
Description
- Color-coded to avoid confusion
- Made of soda-lime glass
- DE-M marked according to the German calibration law
- Calibrated to deliver, 5 second waiting time
- Inclusive batch certificate
- Brown graduation
Specifications
Yes | |
DIN EN ISO 648 | |
Soda Lime Glass | |
Brown | |
Not listed | |
Volumetric Pipet |
To Deliver | |
AS | |
Yes | |
5 sec. | |
2.5 mL |