Learn More
Witeg™ Class AS Special Sizes Graduated Pipettes
With tolerances according to DIN EN ISO 648.
63.15€
Specifications
Calibrated | Yes |
---|---|
Calibration | To Deliver |
Certifications/Compliance | DIN EN ISO 648 |
Class | AS |
Material | Soda Lime Glass |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
13437577
|
Witeg™
3 535 025 |
2.5 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
- Color-coded to avoid confusion
- Made of soda-lime glass
- DE-M marked according to the German calibration law
- Calibrated to deliver, 5 second waiting time
- Inclusive batch certificate
- Brown graduation
Specifications
Yes | |
DIN EN ISO 648 | |
Soda Lime Glass | |
Brown | |
Volumetric Pipet |
To Deliver | |
AS | |
Yes | |
5 sec. |