missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ COL4A6 (Human) Recombinant Protein

Human COL4A6 full-length ORF ( AAH05305, 1 a.a. - 73 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00001288-P01.25ug

 View more versions of this product

Product Code. 16193371

  • 521.00€ / 25µg
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Sequence: MLINKLWLLLVTLCLTEELAAAGEKSYGKPCGGQDCSGSCQCFPEKGARHNLQLLNDMAGRLYHFSEVLPNLF
Specifications

Specifications

AAH05305
collagen, type IV, alpha 6
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
COL4A6
GST
1288
Wheat germ expression system
25 μg
MGC88184
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.