Learn More
VELP Scientifica™ Condensation Kit for Water Samples
The condensation kit for water samples consists of a 3.5 liters tank and a condenser that permits the condensation and collection of the evaporated liquid without requiring any operator intervention.
Brand:  VELP Scientifica™ A00000477
Product Code. 17824687
  • 554.00€ / Each
Estimated Shipment: 21-05-2025
Description
The condensation kit permits the condensation and collection of the evaporated liquid without operator intervention.
  • Consists of a 3.5 liter tank
  • For low nitrogen content samples, such as water samples, the TKN analysis requires a high sample volume.
  • During the digestion step, the evaporation of the liquid could lead to the filling of the KS 1000 dilution tank and a relevant increase in the reagents’ temperature.
  • Required when the sum of the sample volumes to be digested is more than 500 mL. 
  • Can be used in substituion of the dilution tank or it can be used as an additional stage.
Specifications
Specifications
Condensation Kit
Product Title
VELP Scientifica™ Condensation Kit for Water Samples > Velp Scientifica™ Consendation Kit