Learn More
JG Finneran Associates™ Conical Inserts
Use these inserts for a variety of applications. JG Finneran™ Conical Inserts feature a polymer spring which acts as a shock absorber and protects against breakage if the needle bottoms out.
Brand:  JG Finneran Associates™ 4005BS-625
 View more versions of this product
Product Code. 10537352
  • 63.85€ / Pack of 100
Estimated Shipment: 07-07-2025
Description
  • Available preassembled with a polymer bottom spring or patented Top Spring™
  • Polymer spring acts as a shock absorber that protects against breakage if the needle bottoms out
  • Conical design permits complete sample removal
Specifications
Specifications
Glass
Bottom Spring
100 Pk.
50 μL
Big mouth, limited volume, precision formed mandrel
Certificates
Promotions
Promotions
Product Content Correction
Product Title
JG Finneran Associates™ Conical Inserts > Capacity: 50μL; For Use With Big mouth, limited volume, precision formed mandrel; With Bottom Spring