missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Connexin 43/GJA1 Recombinant Protein Antigen
Recombinant Protein
Brand: Novus Biologicals™ NBP2-68678PEP
This item is not returnable.
View return policy
Specifications
Antibody Competition | |
2697 | |
100 μL | |
connexin 43, connexin-43, C x 43, C x 43 gap junction protein, alpha-like, DFNB38, Gap junction 43 kDa heart protein, gap junction alpha-1 protein, gap junction protein, alpha 1, 43 kDa, gap junction protein, alpha 1, 43 kDa (connexin 43), GJAL, HSS, ODD, ODDD, ODOD, SDTY3 | |
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-68678. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL | |
>80% by SDS-PAGE and Coomassie blue staining |
PBS and 1 M Urea, pH 7.4. | |
MolecularWeight-theroretical: 24 kDa | |
Store at -20°C. Avoid freeze/thaw cycles. | |
GJA1 | |
Cancer, Cellular Markers, Core ESC Like Genes, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
Connexin 43/GJA1 |