Learn More
Sperian™ Honeywell™ Cotton/Elasthane White Gloves
For handlings requiring a clean glove without any plush. Sperian™ Honeywell™ Cotton/Elasthane White Gloves are excellent for finishing operations in mechanical, clock-making, jewelry and photography.
Brand: Sperian™ RU711_8
Description
- Industrial cut, edged border
- Ambidextrous and thus more cost-effective
- Cotton interlock ensures the glove's mechanical performance while absorbing hand perspiration for added comfort
- Tight knit gives the glove perfect support and limits the penetration of dirt
- Ribbed at the wrist to ensure a good level of wrist comfort and eliminates the risk of it getting caught
Specifications
Class 5 | |
Ambidextrous | |
Gloves | |
Finishing operations in mechanical, clock-making, jewelery, photography. Handlings requiring a clean glove without any plush. | |
19.5-21 cm |
Jersey Knit | |
Cotton, Elasthane | |
8/10 | |
12/Pk. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 13
Product Content Correction
Product Title
Sperian™ Honeywell™ Cotton/Elasthane White Gloves > Size: 8/10