missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP1-54775
386.40 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
CPS1 Polyclonal specifically detects CPS1 in Human, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
CPS1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
carbamoyl-phosphate synthase [ammonia], mitochondrial, carbamoyl-phosphate synthase 1, mitochondrial, carbamoyl-phosphate synthetase 1, mitochondrial, carbamoylphosphate synthetase I, Carbamoyl-phosphate synthetase I, CPSase I, CPSASE1, EC 6.3.4.16 | |
Rabbit | |
165 kDa | |
100 μL | |
Primary | |
Yeast: 77%. | |
Human, Pig, Rat, Bovine, Canine, Canine, Equine, Guinea Pig, Rabbit, Yeast | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
P31327 | |
CPS1 | |
Synthetic peptides corresponding to CPS1(carbamoyl-phosphate synthetase 1, mitochondrial) The peptide sequence was selected from the middle region of CPS1. Peptide sequence YPSVTNYLYVTYNGQEHDVNFDDHGMMVLGCGPYHIGSSVEFDWCAVSSI. | |
Protein A purified | |
RUO | |
1373 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CPS1 Antibody, Novus Biologicals™ > 100 ul; Unconjugated
Spot an opportunity for improvement?Share a Content Correction