missing translation for 'onlineSavingsMsg'
Learn More

CPS1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

483.00€

Specifications

Antigen CPS1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18263261
View Documents
Bio-Techne
NBP1-54775
100 ÎĽL
483.00€
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CPS1 Polyclonal specifically detects CPS1 in Human, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

CPS1
Polyclonal
Purified
RUO
carbamoyl-phosphate synthase [ammonia], mitochondrial, carbamoyl-phosphate synthase 1, mitochondrial, carbamoyl-phosphate synthetase 1, mitochondrial, carbamoylphosphate synthetase I, Carbamoyl-phosphate synthetase I, CPSase I, CPSASE1, EC 6.3.4.16
CPS1
IgG
Protein A purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
P31327
1373
Synthetic peptides corresponding to CPS1(carbamoyl-phosphate synthetase 1, mitochondrial) The peptide sequence was selected from the middle region of CPS1. Peptide sequence YPSVTNYLYVTYNGQEHDVNFDDHGMMVLGCGPYHIGSSVEFDWCAVSSI.
Primary
165 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CPS1 Antibody, Novus Biologicals™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.