Learn More
Afora™ Cylindrical Addition Funnel
Add or transfer liquids while controlling flow. Afora™ Cylindrical Addition Funnel includes a PTFE stopcock.
Brand:  Afora™ 1071/E
 View more versions of this product
Product Code. 11946338
  • 182.00€ / Each
Estimated Shipment: 12-03-2025
Description
  • Manufactured from blown glass
  • Includes PTFE stopcock for flow control
  • Cylindrical shape
  • Ground glass joints
Specifications
Specifications
56 mm
Glass
No
29/32
Dropping Funnel
250 mL
Plastic closure
Plastic cap, stopcock
PTFE