Learn More
Afora™ Cylindrical Addition Funnel
Add or transfer liquids while controlling flow. Afora™ Cylindrical Addition Funnel includes a PTFE stopcock.
176.00€ - 182.00€
Specifications
Material Glass
Description Plastic closure
Graduated No
Includes Plastic cap, stopcock
Joint Number 29/32
View More Specs
Products 2
Product Code Brand Capacity (Metric) Diameter (Metric)Top Joint Number Price Quantity & Availability  
Product Code Brand Capacity (Metric) Diameter (Metric)Top Joint Number Price Quantity & Availability  
11966328
View Documents
Afora™
1070/E
100 mL 36 mm 29/32
176.00€
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
11946338
View Documents
Afora™
1071/E
250 mL 56 mm 29/32
182.00€
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
Description
Description
  • Manufactured from blown glass
  • Includes PTFE stopcock for flow control
  • Cylindrical shape
  • Ground glass joints
Specifications
Specifications
Glass
No
29/32
Dropping Funnel
Plastic closure
Plastic cap, stopcock
PTFE