missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CyTRAK Orange™
334.00€ - 621.00€
Specifications
Content And Storage | Store at 4°C in the dark. Do not freeze. |
---|---|
For Use With (Application) | Flow Cytometry, Immunocytochemistry/Immunofluorescence, Live Imaging Microscopy |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18054234
|
Novus Biologicals™
NBP2-81127-50UL |
50 μL |
334.00€
50µL |
Estimated Shipment: 28-11-2024
Log in to see stock. |
|||||
18277318
|
Novus Biologicals™
NBP2-81127-200UL |
200 μL |
621.00€
200µL |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
Specifications
Store at 4°C in the dark. Do not freeze. | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence, Live Imaging Microscopy |