Learn More
Afora™ Decanting Funnel with Cane
Separates immiscible, heterogenous liquids. Afora™ Decanting Funnel with Cane includes a plastic stopper and Rotaflo TF wrench.
Brand:  Afora™ 1099/TFE
 View more versions of this product
Product Code. 11931109
  • 298.00€ / Each
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Pear-shaped body
  • Cone-shaped bottom
  • Glass with plastic stopcock
  • Includes Rotaflo TF wrench
Specifications
Specifications
152 mm
2000 mL
Glass
No