Learn More
Afora™ Decanting Funnel with Cane
Separates immiscible, heterogenous liquids. Afora™ Decanting Funnel with Cane includes a plastic stopper and Rotaflo TF wrench.
144.00€ - 298.00€
Specifications
Material Glass
Graduated No
View More Specs
Products 4
Product Code Brand Capacity (Metric) Diameter (Metric)Top Price Quantity & Availability  
Product Code Brand Capacity (Metric) Diameter (Metric)Top Price Quantity & Availability  
11921099
View Documents
Afora™
1098/TFE
1000 mL 122 mm
193.00€
Each
Estimated Shipment: 04-04-2025
to see stock.
Add to basket
11926428
View Documents
Afora™
1096/TFE
250 mL 80 mm
144.00€
Each
Estimated Shipment: 04-04-2025
to see stock.
Add to basket
11971079
View Documents
Afora™
1097/TFE
500 mL 98 mm
180.00€
Each
Estimated Shipment: 04-04-2025
to see stock.
Add to basket
11931109
View Documents
Afora™
1099/TFE
2000 mL 152 mm
298.00€
Each
Estimated Shipment: 04-04-2025
to see stock.
Add to basket
Description
Description
  • Pear-shaped body
  • Cone-shaped bottom
  • Glass with plastic stopcock
  • Includes Rotaflo TF wrench
Specifications
Specifications
Glass
No